Ammoniumheptamolybdat Tetrahydrat, weißes kristallines Pulver ACS

Lieferant: MP Biomedicals
Achtung

0215249780 0215249790 0215249791
ICNA0215249780EA 59 EUR
ICNA0215249780 ICNA0215249790 ICNA0215249791
Ammoniumheptamolybdat Tetrahydrat, weißes kristallines Pulver ACS
Ammoniumheptamolybdat Tetrahydrat

Ammonium Molybdate Tetrahydrate is an analgesic.


Ammonium Molybdate Tetrahydrate is an analgesic.


Bitte beachten Research Use Only (RUO).

Formel: (NH₄)₆Mo₇O₂₄·4H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00167059
CAS-Nummer: 12054-85-2

Bestellen

Spezifikation Testergebnisse

Assay (as MoO3) 81,0 - 83,0%
Heavy Metals (as Pb) ≤0,001%
Insoluble Matter ≤0,005%
Chloride (Cl) ≤0,002%
Arsenate, Phosphate and Silicate (as SiO2) ≤0,001%
Phosphate (PO4) ≤5 ppm
Sulphate (SO4) ≤0,02%
Magnesium ≤,005%
Potassium ≤0,002%
Sodium ≤0,01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR