Natriumdiphenylamin-4-sulfonat, Pulver ACS Redoxindikator

Lieferant: MP Biomedicals
Achtung

Synonyme: 4-(Phenylamino)benzolsulfonsäure Natriumsalz, Diphenylamin-4-sulfonsäure Natriumsalz, Natrium(4-anilinobenzolsulfonat), Natrium-N-phenylsulfanilat

0215251905
ICNA0215251905EA 71.1 EUR
ICNA0215251905
Natriumdiphenylamin-4-sulfonat, Pulver ACS Redoxindikator
Natriumdiphenylamin-4-sulfonat

Diphenylamine-4-sulfonic acid sodium salt is a redox-indicator.


Soluble in water (50 mg/ml - hazy dark yellow-green solution).


Bitte beachten Research Use Only (RUO).

Formel: C₁₂H₁₀NNaO₃S
Molecular Weight: 271,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036468
CAS-Nummer: 6152-67-6

Bestellen

Spezifikation Testergebnisse

Sensitivity as Indicator Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR