Molybdatophosphorsäure Hydrat, Pulver ACS

Lieferant: MP Biomedicals
Gefahr

Synonyme: Phosphormolybdänsäure Hydrat, Dodecamolybdophosphorsäure Hydrat

0215255605
ICNA0215255605EA 54.2 EUR
ICNA0215255605
Molybdatophosphorsäure Hydrat, Pulver ACS
Molybdatophosphorsäure Hydrat

Phosphomolybdic acid hydrate is employed in preparation of a hybrid organic-inorganic, electroactive material with potential application as a cation-insertion electrode in batteries.


Very soluble in alcohol, ether; slightly soluble in less than 0,4 part water.


Bitte beachten Research Use Only (RUO).

Formel: H₃[P(Mo₃O₁₀)₄]·nH₂O
Molecular Weight: 1843,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149913
CAS-Nummer: 51429-74-4
UN: 3260
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Insoluble Matter ≤0.01%
Chloride (Cl) <0,02%
Sulphate (SO4) <0,025%
Ammonium (NH4) 0,01%
Calcium (Ca) 0,02%
Heavy Metals (as Pb) <0,005%
Iron (Fe) <0,005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR