Acid blue 29, schwarzes Pulver C.I. 20460

Lieferant: MP Biomedicals

Synonyme: Disodium 4-amino-5-hydroxy-3-((3-nitrophenyl)azo)-6-(phenylazo)naphthalene-2,7-disulphonate, C.I. 20460, Mordant Blue 82

0215259625
ICNA0215259625EA 62.1 EUR
ICNA0215259625
Acid blue 29, schwarzes Pulver C.I. 20460
Acid blue 29

Acid Blue 29 is an anionic disazo dye.


Slightly soluble in methyl cellosolve and water (20 mg/ml); very slightly soluble in ethanol (2 mg/ml).


Bitte beachten Research Use Only (RUO).

Formel: C₂₂H₁₄N₆O₉S₂Na₂
Molecular Weight: 616,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009960
CAS-Nummer: 5850-35-1

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
UV/Vis: λmax (water) 600 - 605 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR