Carmin (Calcium-Aluminium-Verlackung von Carminsäure), rotes Pulver C.I. 75470

Lieferant: MP Biomedicals

Synonyme: CI 75470

0215263425
ICNA0215263425EA 210 EUR
ICNA0215263425
Carmin (Calcium-Aluminium-Verlackung von Carminsäure), rotes Pulver C.I. 75470
Carmin (Calcium-Aluminium-Verlackung von Carminsäure)

Soluble in ethanol, alkali hydroxides, borax (0,8 mg/ ml), ammonium hydroxide (1 mg/ml), water (8 mg/ml) or ethanol (0,8 mg/ml). The addition of an acid or base can help increase the water solubility. Partly soluble in hot water; practically insoluble in cold water (8 mg/ ml) or dilute acids.


Soluble in ethanol, alkali hydroxides, borax (0,8 mg/ ml), ammonium hydroxide (1 mg/ml), water (8 mg/ml) or ethanol (0,8 mg/ml). The addition of an acid or base can help increase the water solubility. Partly soluble in hot water; practically insoluble in cold water (8 mg/ ml) or dilute acids.

Formel: C₂₂H₂₀O₁₃
Molecular Weight: 492,389 g/mol
MDL: MFCD00167028
CAS-Nummer: 1390-65-4
EINECS: 215-724-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR