Indigocarmin, dunkles Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
Acid Blue 74, Food blue No. 2
0215263805
0215263880
ICNA0215263805EA
30.5
EUR
ICNA0215263805
ICNA0215263880
Indigocarmin, dunkles Pulver
Indigocarmin
Indigo Carmine is a blue, anionic indigoid dye.
Indigo Carmine is useful in the staining of Negri bodies. It can be used as a contrast stain for plasma when red nuclear stains are used.
- Presentation: Dark Powder
- Soluble in water (3 mg/mL) and ethanol (3 mg/mL).
- Storage temperature: Store at room Temperature, protect from light.
Formel:
C₁₆H₈N₂Na₂O₈S₂ Schmelzpunkt: ? 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005723 CAS-Nummer: 860-22-0 EINECS: 212-728-8 Merck Index: 13,04969 |
Spezifikation Testergebnisse
Identity Test | Passes |
UV/Vis: l max (water) | 605-615nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...