Lichtgrün SF gelblich, violettes Pulver C.I. 42095
Lieferant: MP Biomedicals
Synonyme:
Acid Green 5, Dihydrogen(ethyl)[4-[p-[ethyl(m-sulfonatobenzyl)amino]-p'-sulfonatobenzhydrylen]cyclohexa-2,5-dien-1-yliden](m-sulfonatobenzyl)ammonium Dinatriumsalz
0215264125
ICNA0215264125EA
146
EUR
ICNA0215264125
Lichtgrün SF gelblich, violettes Pulver C.I. 42095
Lichtgrün SF gelblich
Acid Green 5 is useful as a general plasma stain, as a histological stain, and a cytological counterstain. Can be use as a cytological counterstain and as a substitute for aniline blue in Gomori′s one-step trichrome procedure.
Soluble in water.
Bitte beachten Research Use Only (RUO).
Formel:
C₃₇H₃₄N₂Na₂O₉S₃ Molecular Weight: 792,86 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012121 CAS-Nummer: 5141-20-8 |
Spezifikation Testergebnisse
Identity Test | Passes |
UV/VIS: λmax (H₂O) | 630 ± 5 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...