Malachitgrün-Oxalat, grünes Pulver C.I. 42000
Lieferant: MP Biomedicals
|
Gefahr
|
Synonyme:
4,4'-Di(dimethylamino)trityliumoxalat
0215264225
0215264280
ICNA0215264225EA
40.4
EUR
ICNA0215264225
ICNA0215264280
Malachitgrün-Oxalat, grünes Pulver C.I. 42000
Malachitgrün-Oxalat
Malachite green is use as counter-stain against fuchsin which stains gram-positive and gram-negative bacteria reddish colors and safranin which stains nuclei red. Malachite green stain background to the surrounding tissue blue-green.
Malachite green is also used as a pH Indicator between pH 0.2 (green) - 1.8 (blue-green). Used as a botanical counterstain.
Soluble in water (40 mg/ ml), EGME (40 mg/ ml), EtOH (50 mg/ ml).
Formel:
C₅₂H₅₄N₄O₁₂ Molecular Weight: 927.02 g/mol Schmelzpunkt: 159 |
MDL:
MFCD00011766 CAS-Nummer: 2437-29-8 UN: 2811 ADR: 6.1 III Merck Index: 13,05722 |
|
Spezifikation Testergebnisse
Identity Test | Passes |
UV/Vis: l max (water) | 614 - 618 nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...