Methylblau

Lieferant: MP Biomedicals

Synonyme: Baumwollblau, Acid Blue 93, Anilinblau WS, Wasserblau 6B, Aniline blue water soluble, Anilinblau wasserlöslich, Aniline blue WS, Dinatrium-[[4-[bis[4-[(sulfonatophenyl)amino]phenyl]methylen]cyclohexa-2,5-dien-1-yliden]amino]benzolsulfonat

0215264325
ICNA0215264325EA 142 EUR
ICNA0215264325
Methylblau
Methylblau
Formel: C₃₇H₂₇N₃Na₂O₉S₃
Molecular Weight: 799,81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058509
CAS-Nummer: 28983-56-4
EINECS: 249-352-9
Merck Index: 13,06055

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR