Methylene Green zinc chloride double salt, dunkelgrünes Pulver C.I. 52020

Lieferant: MP Biomedicals
Achtung

0215264480 0215264405
ICNA0215264480EA 181 EUR
ICNA0215264480 ICNA0215264405
Methylene Green zinc chloride double salt, dunkelgrünes Pulver C.I. 52020
Methylene Green zinc chloride double salt

Dark Green almost Black Fine Powder


Soluble in water (10 mg/ ml-opaque, deep blue solution) and alcohol.

Formel: C₁₆H₁₇ClN₄O₂S·0.5ZnCl₂
MDL: MFCD00151524
CAS-Nummer: 6722-15-2
EINECS: 220-231-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR