Sudan IV, rotbraun, Pulver C.I. 26105
Lieferant: MP Biomedicals
This product is soluble in chloroform (10 mg/ml), toluene, methanol. Saturated solutions can be prepared in isopropanol. Insoluble in water.
Sudan IV has been used industrially to color oils, waxes, shoe polish, greases, oily-resin lacquers, varnishes, cellulose acetate and acrylic resins, and in wood stains. As a biological stain, it is used primarily to stain lipids and fatty substances present in cells and tissues. The color imparted is a function of the solubility of the dye in the substance affected and does not involve chemical bonding. A saturated solution in isopropanol is prepared for use as a fat stain in animal tissue.
Formel:
C₂₄H₂₀N₄O |
MDL:
MFCD00003893 CAS-Nummer: 85-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...