Kupfer(II)nitrat Trihydrat, Pulver

Lieferant: MP Biomedicals

Synonyme: Kupferdinitrat Trihydrat

0215350180 0215350190
ICNA0215350180EA 34.5 EUR
ICNA0215350180 ICNA0215350190
Kupfer(II)nitrat Trihydrat, Pulver
Kupfer(II)nitrat Trihydrat
Formel: Cu(NO₃)₂·3H₂O
MDL: MFCD00149671
CAS-Nummer: 10031-43-3
UN: 1477
ADR: 5.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR