Calciumchlorid, wasserfrei ≥95%, weißes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: Chlorcalcium

0215350291 0215350280 0215350290
ICNA0215350291EA 140 EUR
ICNA0215350291 ICNA0215350280 ICNA0215350290
Calciumchlorid, wasserfrei ≥95%, weißes Pulver
Calciumchlorid

Anhydrous Calcium Chloride is highly hygroscopic and is often used as a desiccant. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability.


Soluble in water with heat liberation, ethanol, acetic acid, acetone.

Formel: CaCl₂
Siedepunkt: 1600 °C (1013 hPa)
Schmelzpunkt: 772 °C
Dichte: 2,174 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010903
CAS-Nummer: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR