Serotonin Hydrochlorid, blassgraues Pulver

Lieferant: MP Biomedicals
Achtung

0215376591 0215376525 0215376550 0215376580
ICNA0215376591EA 803 EUR
ICNA0215376591 ICNA0215376525 ICNA0215376550 ICNA0215376580
Serotonin Hydrochlorid, blassgraues Pulver
Serotonin Hydrochlorid

A tryptophan based amino acid neurotransmitter.


Serotonin hydrochloride is used as a neurotransmitter and functions as an endogenous serotonin receptor agonist in various biochemical processes. It is an active component, which is used to regulate the mood, emesis, cognition and appetite in vivo.


This neurotransmitter is located in the central nervous system and functions as an endogenous SR (serotonin receptor) agonist in a variety of biochemical processes. In the majority of the human bodies, serotonin is located in enterochromaffin ("Kulchitsky cells"). Serotonin is found in the gastrointestinal tract and platelets. Furthermore, Serotonin functions as a growth factor in some cells and facilitates wound healing in conjunction with platelets. Serotonin is thought to be a key component in the regulation of moods, cognition, and personality traits. As a result, this compound is essential in studying the variations in these traits.


Soluble in water: 100 mg/ml; 0.1 M HCl (22 mg/ml); moderately soluble in ethanol (5.3 mg/ml); insoluble in ether, chloroform; unstable in aqueous base.


Bitte beachten Store Under Nitrogen.

Formel: C₁₀H₁₃ClN₂O
Lagertemperatur: Gefrierschrank
MDL: MFCD00012686
CAS-Nummer: 153-98-0
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard Loss on Drying
Loss on Drying ≤1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR