1,3,5-Tri-O-benzoyl-α-D-ribofuranose, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: 1,3,5-Tri-O-benzoyl-α-D-ribofuranose, 1,3,5-Tri-O-benzoyl-a-D-ribofuranose

0215396801
ICNA0215396801EA 31 EUR
ICNA0215396801
1,3,5-Tri-O-benzoyl-α-D-ribofuranose, weißes Pulver
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose

Starting material for nucleosides.


Soluble in CHCl₃ (50 mg/ml-clear, colorless solution)

Formel: C₂₆H₂₂O₈
Molecular Weight: 462,45 g/mol
MDL: MFCD00080818
CAS-Nummer: 22224-41-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Melting Point 132 - 142 °C
Optical Rotation (c=1, CHCl₃) +85° ± 5°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR