Azomethine H monosodium salt monohydrate, powder_orange
Lieferant: MP Biomedicals
Synonyme:
4-Hydroxy-5-(2-hydroxybenzylideneamino)naphthalene-2,7-disulphonic acid monosodium salt
0215416905
0215416901
0215416910
ICNA0215416905EA
154
EUR
ICNA0215416905
ICNA0215416901
ICNA0215416910
Azomethine H monosodium salt monohydrate, powder_orange
Azomethine H monosodium salt monohydrate
Azomethine H is a reagent for the determination of boron in soil, plants, composts, manure, water and nutrient solutions.
Azomethine H is a reagent for the determination of boron in soil, plants, composts, manure, water and nutrient solutions.
Soluble in hot or cold water (0,1 w/v%-clear).
Formel:
C₁₇H₁₂NNaO₈S₂·H₂O Molecular Weight: 463.41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149588 CAS-Nummer: 304655-82-1 EINECS: 250-975-3 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Reference |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...