Cefazolin Natriumsalz

Lieferant: MP Biomedicals
Gefahr

Synonyme: Sodium (6R,7R)-7-(2-(1H-tetrazol-1-yl)acetamido)-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

0215494680 0215494690 0215494691
ICNA0215494680EA 70.2 EUR
ICNA0215494680 ICNA0215494690 ICNA0215494691
Cefazolin Natriumsalz
Cefazolin sodium salt

Storage temperature.: +4 °C

Formel: C₁₄H₁₄N₈NaO₄S₃
MDL: MFCD00056883
CAS-Nummer: 27164-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR