Kojisäure, weißes kristallines Pulver
Lieferant: MP Biomedicals
0215515710
0215515725
0215515701
0215515705
ICNA0215515710EA
230
EUR
ICNA0215515710
ICNA0215515725
ICNA0215515701
ICNA0215515705
Kojisäure, weißes kristallines Pulver
Kojisäure
Kojic acid is used as an antibiotic reagent. It is tyrosinase inhibitor. Kojic acid is a chelation agent.
Soluble in water, ethanol, acetone; Slightly soluble in ethyl acetate, chloroform, pyridine.
Formel:
C₆H₆O₄ Siedepunkt: 323 °C (1013 hPa) Schmelzpunkt: 148…157 °C Dichte: 1,58 g/cm³ (20 °C) |
MDL:
MFCD00006580 CAS-Nummer: 501-30-4 EINECS: 207-922-4 Merck Index: 13,05334 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...