Kojisäure, weißes kristallines Pulver

Lieferant: MP Biomedicals

0215515710 0215515725 0215515701 0215515705
ICNA0215515710EA 230 EUR
ICNA0215515710 ICNA0215515725 ICNA0215515701 ICNA0215515705
Kojisäure, weißes kristallines Pulver
Kojisäure

Kojic acid is used as an antibiotic reagent. It is tyrosinase inhibitor. Kojic acid is a chelation agent.


Soluble in water, ethanol, acetone; Slightly soluble in ethyl acetate, chloroform, pyridine.

Formel: C₆H₆O₄
Siedepunkt: 323 °C (1013 hPa)
Schmelzpunkt: 148…157 °C
Dichte: 1,58 g/cm³ (20 °C)
MDL: MFCD00006580
CAS-Nummer: 501-30-4
EINECS: 207-922-4
Merck Index: 13,05334

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR