Mangan(II)sulfat Monohydrat, powder_pale pink

Lieferant: MP Biomedicals
Achtung

Synonyme: Mangansulfat Monohydrat

0215533690 0215533691
ICNA0215533690EA 63.9 EUR
ICNA0215533690 ICNA0215533691
Mangan(II)sulfat Monohydrat, powder_pale pink
Mangan(II)sulfat Monohydrat

Magnesium sulphate is used as a colorant in dyes and red glazes for porcelain. As a trace essential element it is a nutritional factor for all animals; often a supplement in feed. It is used in biological assays to study the effects of manganese on the central nervous system, and to assess exposure limits.


Magnesium sulphate is used as a colorant in dyes and red glazes for porcelain. As a trace essential element it is a nutritional factor for all animals; often a supplement in feed. It is used in biological assays to study the effects of manganese on the central nervous system, and to assess exposure limits.


Soluble in water (77 mg/ml), yielding a clear, light pink solution (Lit.).

Formel: MnSO₄·1H₂O
Molecular Weight: 169.02 g/mol
Schmelzpunkt: 700
MDL: MFCD00149159
CAS-Nummer: 10034-96-5
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR