Methyl myristoleate ≥99%, Flüssigkeit

Lieferant: MP Biomedicals

Synonyme: Myristoleic Acid Methyl Ester, Methyl cis-9-tetradecenoate

0215576150 0215576180 0215576190
ICNA0215576150EA 70.2 EUR
ICNA0215576150 ICNA0215576180 ICNA0215576190
Methyl myristoleate ≥99%, Flüssigkeit
Methyl myristoleate

Sealed ampule

Formel: C₁₅H₂₈O₂
Molecular Weight: 240.387 g/mol
MDL: MFCD00056299
CAS-Nummer: 56219-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR