2-(4-Nitrophenyl)ethanol, Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol
0215589005
0215589050
0215589010
ICNA0215589005EA
61.2
EUR
ICNA0215589005
ICNA0215589050
ICNA0215589010
2-(4-Nitrophenyl)ethanol, Pulver
2-(4-Nitrophenyl)ethanol
p-Nitrophenethyl alcohol is used as a protecting reagent for phosphates and O6 of deoxyguanosine.
Soluble in acetone (50 mg/ml) and methanol (25 mg/ml).
Formel:
C₈H₉NO₃ Schmelzpunkt: 59…62 °C (Lit.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00010202 CAS-Nummer: 100-27-6 EINECS: 202-835-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...