p-Nitrophenyl-N-acetyl-α-D-glucosaminide, weißes Pulver

Lieferant: MP Biomedicals

0215590180
ICNA0215590180EA 811 EUR
ICNA0215590180
p-Nitrophenyl-N-acetyl-α-D-glucosaminide, weißes Pulver
p-Nitrophenyl-N-acetyl-α-D-glucosaminide
Formel: C₁₄H₁₈N₂O₈
Molecular Weight: 342.304 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 10139-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR