Paromomycin sulphate ≥97%, Pulver

Lieferant: MP Biomedicals

Synonyme: (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate(1:x)

0215605625
ICNA0215605625EA 593 EUR
ICNA0215605625
Paromomycin sulphate ≥97%, Pulver
Paromomycin sulphate

Paromomycin is a broad spectrum aminoglycoside antibiotic produced by Streptomyces rimosus var. paromomycinus. It has in vitro and in vivo activity similar to neomycin. It is effective against gram-negative bacteria, gram-positive bacteria, some protozoan species, and limited anti helminth. It is used to study bacterial protein synthesis at the level of 16S ribosomal RNA and 30S ribosome assembly. Paromomycin is used to study cytosine-cytosine (CC) mismatch-containing RNA molecules and is used to inhibit cryptosporidium infection of a human enterocyte cell line.


Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.


Soluble in water (25% w/v - light opalescent solution).

Formel: C₂₃H₄₅N₅O₁₄·H₂SO₄
Molecular Weight: 615,6 g/mol
MDL: MFCD00079278
CAS-Nummer: 1263-89-4

Bestellen

Spezifikation Testergebnisse

Form White Powder
Storage Store at Room Temperature (15 - 30 °C)
Potency 675 μg/mg
Typical Working Concentration 100 mg/L

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR