2-Phenylethanol, Flüssigkeit

Lieferant: MP Biomedicals
Gefahr

Synonyme: 2-Phenylethylalkohol, β-Phenylethylalkohol, Phenylethylalkohol, Benzolthanol, β-Hydroxyethylbenzol, Benzylcarbinol, Phenethylalkohol

0215617691
ICNA0215617691EA 85.4 EUR
ICNA0215617691
2-Phenylethanol, Flüssigkeit
2-Phenylethanol

2-Phenylethanol, an alcohol potentially used as an anti-microbial.


Soluble in most fixed oils, common organic solvents, water and propylene glycol.

Formel: C₆H₅CH₂CH₂OH
Molecular Weight: 122,17 g/mol
Schmelzpunkt: –27 °C
MDL: MFCD00002886
CAS-Nummer: 60-12-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR