Piperacillin sodium salt, weißes Pulver

Lieferant: MP Biomedicals
Gefahr

Synonyme: Piperacillin sodium, Sodium (2S,5R,6R)-6-((R)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

0215626801 0215626805 0215626810
ICNA0215626801EA 105 EUR
ICNA0215626801 ICNA0215626805 ICNA0215626810
Piperacillin sodium salt, weißes Pulver
Piperacillin sodium salt

Piperacillin is a semisynthetic, broad-spectrum ureidopenicillin antibiotic. It is derived from ampicillin. Piperacillin inhibits the last stage of bacterial cell wall synthesis by binding to certain penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes. Piperacillin may interfere with autolysin inhibitors.
It has been used in pharmacokinetic studies in order to optimize antimicrobial therapy in patients with sepsis. It is used to study piperacillin hypersensitivity reactions and to study multidrug-resistant organisms.
Piperacillin inhibits the last stage of bacterial cell wall synthesis by binding to certain penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes. Piperacillin may interfere with autolysin inhibitors.


  • Presentation: White Powder
  • Soluble in aqueous solution (10 mg/mL-clear, colorless solution), in water (50 mg/mL-clear colorless to faint yellow solution).
  • Storage temperature: Store at + 4 °C

Formel: C₂₃H₂₇N₅O₇S
Lagertemperatur: Kühlschrank
MDL: MFCD00056867
CAS-Nummer: 59703-84-3

Bestellen

Spezifikation Testergebnisse

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR