(-)-Chinin Hydrochlorid Dihydrat, weißes Pulver

Lieferant: MP Biomedicals
Achtung

0215648650 0215648605 0215648625 0215648680
ICNA0215648650EA 161 EUR
ICNA0215648650 ICNA0215648605 ICNA0215648625 ICNA0215648680
(-)-Chinin Hydrochlorid Dihydrat, weißes Pulver
(-)-Chinin Hydrochlorid Dihydrat

Quinine Hydrochloride is an alkaloid originally isolated from the Cinchona family of South American trees. It is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent.


Quinine is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent; also used as a resolving agent. A potassium channel blocker. Also used as a flavoring agent in carbonated beverages, a skeletal muscle relaxant and an antiprotozoal agent for fish. A chiral catalyst used in highly enantioselective [2+2] cycloadditions. A resolving agent for carboxylic acids. Catalyzes the kinetic resolution of furanones.


Potassium channel blocker.


Soluble in water (1 g/16 ml at 25 °C; 1 g/0,5 ml at 100 °C), ethanol (1 g/1 ml), glycerol (~1 g/7 ml), chloroform (~1 g/ml), or ether (1 g/350 ml).

Formel: C₂₀H₂₅ClN₂O₂·2H₂O
Flammpunkt: 122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151248
CAS-Nummer: 6119-47-7
EINECS: 205-001-1

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR