Sudanblau II, dunkel rot/violett fast schwarzes Pulver C.I. 61554

Lieferant: MP Biomedicals

Synonyme: Sudan Blue 2, 1,4-Bis(butylamino)-9,10-anthraquinone, 1,4-bis(butylamino)anthracene-9,10-dione, 1,4-Bis(butylamino)anthracene-9,10-dione, Sudan Blue II

0215663550 0215663580
ICNA0215663550EA 118 EUR
ICNA0215663550 ICNA0215663580
Sudanblau II, dunkel rot/violett fast schwarzes Pulver C.I. 61554
Sudanblau II

Solvent Blue 35 is a member of the Sudan family of hydrophobic fat-staining dyes. In structural classification it is a diazo dyes.
Solvent Blue 35 is predominantly used for staining triglycerides in animal tissues (frozen sections). With the use of certain solvents, may also be used to stain some protein bound lipids in paraffin sections.


  • Presentation: Dark Red/Purple almost Black Powder
  • Dye content: ∼ 90%
  • Soluble in chloroform(4mg/mL).
  • UV/Visible Absorbance: λmax: 604 nm (2nd), λmax: 652 nm

Formel: C₂₂H₂₆N₂O₂
Schmelzpunkt: 120…122 °C (Lit.)
Dichte: 1,23 g/mL (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011714
CAS-Nummer: 17354-14-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR