Sulfamethoxypyridazin, Pulver
Lieferant: MP Biomedicals
Synonyme:
4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulphonamide
0215671225
ICNA0215671225EA
89.8
EUR
ICNA0215671225
Sulfamethoxypyridazin, Pulver
Sulfamethoxypyridazin
Formel:
C₁₁H₁₂N₄O₃S Molecular Weight: 280,3 g/mol |
MDL:
MFCD00057372 CAS-Nummer: 80-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...