ɑ-Amino-γ-butyrolactone hydrobromide, weißgraues Puder

Lieferant: MP Biomedicals

Synonyme: DL-Homoserine lactone hydrobromide

0215738905
ICNA0215738905EA 96.4 EUR
ICNA0215738905
ɑ-Amino-γ-butyrolactone hydrobromide, weißgraues Puder
alpha-Amino-gamma-butyrolactone hydrobromide

Soluble 5% in water giving colorless to light yellow solution.


Soluble 5% in water giving colorless to light yellow solution.

Formel: C₄H₇NO₂
Molecular Weight: 182,02 g/mol
MDL: MFCD00012723
CAS-Nummer: 6305-38-0

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Melting Point 219 - 226°C
Optical Rotation ~0° (c=20, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR