Patton-Reeder-Reagenz (Calconcarbonsäure), Pulver

Lieferant: MP Biomedicals

Synonyme: 2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthincarbonsäure, Cal-Red®, 2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalin-3-carbonsäure, 3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalin-2-carbonsäure

0215745105
ICNA0215745105EA 50.7 EUR
ICNA0215745105
Patton-Reeder-Reagenz (Calconcarbonsäure), Pulver
Patton-Reeder-Reagenz (Calconcarbonsäure)

2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acid is a useful indicator for calcium titrations with EDTA in the presence of magnesium.


Soluble in methanol (clear reddish violet solution), 50 mg plus 5 ml of 1 M NaOH; slightly soluble in water.

Formel: C₂₁H₁₄N₂O₇S
Molecular Weight: 438,42 g/mol
MDL: MFCD00004078
CAS-Nummer: 3737-95-9

Bestellen

Spezifikation Testergebnisse

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR