Meldola's Blue, schwarzes Pulver C.I. 51175
Lieferant: MP Biomedicals
0215778805
ICNA0215778805EA
191
EUR
ICNA0215778805
Meldola's Blue, schwarzes Pulver C.I. 51175
Meldola's Blue
A dye used in biosensors
Soluble in water (blue violet) alcohol (blue) forms a brown ppt with NaoH in water: Some insolubles in 10 mg/ml ethanol, dark blue to dark purple solution.
Formel:
C₁₈H₁₅ClN₂O·Cl₂Zn Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012113 CAS-Nummer: 7057-57-0 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Reference |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...