Ethylvanillin, weißes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: 3-Ethoxy-4-hydroxybenzaldehyd

0215795980
ICNA0215795980EA 27.3 EUR
ICNA0215795980
Ethylvanillin, weißes Pulver
Ethylvanillin

Ethylvanillin (3-Ethoxy-4-hydroxybenzaldehyde) was used as an internal standard in the determination of phenolic compounds obtained from (alkaline) CuO-based oxidation of humic acid.

Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00006944
CAS-Nummer: 121-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR