Benzocain (Ethyl-4-aminobenzoat), powder_crystalline_white
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
4-Aminobenzoesäureethylester, Äthoform
0215796790
0215796780
ICNA0215796790EA
569
EUR
ICNA0215796790
ICNA0215796780
Benzocain (Ethyl-4-aminobenzoat), powder_crystalline_white
Benzocain (Ethyl-4-aminobenzoat)
Formel:
C₉H₁₁NO₂ Molecular Weight: 165.19 g/mol Siedepunkt: 311 Schmelzpunkt: 88…93 °C Flammpunkt: 112 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007892 CAS-Nummer: 94-09-7 EINECS: 202-303-5 Merck Index: 13,01087 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...