Ethylviolett, dunkles Pulver C.I. 42600

Lieferant: MP Biomedicals

0215801625
ICNA0215801625EA 68.5 EUR
ICNA0215801625
Ethylviolett, dunkles Pulver C.I. 42600
Ethyl violet

Ethyl violet is a cationic triarylmethane dye that is the ethyl homolog of crystal violet. It is a general histological and pathological stain.


  • Visual transition interval: from pH 0,0 (yellow) to pH 3.5 (blue)
  • Soluble in ethanol and water (1 mg/ml)


Ethyl violet has been used to stain pancreatic tissue and elastin, in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. Ethyl violet induces mitochondrial permeability. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.


Soluble in ethanol and water (1 mg/ml).


Visual Transition Interval: From pH 0.0 (yellow) to pH 3.5 (blue)

Formel: C₃₁H₄₂ClN₃
MDL: MFCD00011765
CAS-Nummer: 2390-59-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
UV/Vis: l max (water) 593 - 596 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR