Eisen(III)sulfat Hydrat, blassgraues Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: Dieisentrissulfat Hydrat

0215804283 0215804290 0215804291
ICNA0215804283EA 37.7 EUR
ICNA0215804283 ICNA0215804290 ICNA0215804291
Eisen(III)sulfat Hydrat, blassgraues Pulver
Eisen(III)sulfat Hydrat

Ferric sulphate is used in dyeing as a mordant, and as a coagulant for industrial wastes. It is also used in pigments, and in pickling baths for aluminum and steel. Medically it is used as an astringent and styptic. It also acts as an important oxidative agent.


Soluble in water; sparingly soluble in alcohol; practically insoluble in acetone, ethyl acetate.

Formel: Fe₂(SO₄)₃·xH₂O
Schmelzpunkt: 1539 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149714
CAS-Nummer: 15244-10-7

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Moisture Record Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR