Viktoriablau B, dunkelbraunes Pulver C.I. 44045
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
Basic blue 26, N-(4-((4-(Dimethylamino)phenyl)(4-(phenylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride
Victoria Blue B is used as a nuclear stain in cytology/histochemical applications. Victoria Blue B may be used in the quantitative determination of phospholipids.
Victoria Blue B is used as a nuclear stain in cytology/histochemical applications. Victoria Blue B may be used in the quantitative determination of phospholipids.
More soluble in the lower aliphatic alcohols; slightly soluble in water.
Bitte beachten Research Use Only (RUO).
Formel:
C₃₃H₃₂ClN₃ Molecular Weight: 506,803 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011878 CAS-Nummer: 2580-56-5 EINECS: 219-943-6 |
Spezifikation Testergebnisse
Identity Test | Passes |
UV/Vis: λmax (ethanol) | Report Resut |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...