Zincon sodium salt, braunes Pulver

Lieferant: MP Biomedicals
Achtung

0215831701 0215831705
ICNA0215831701EA 51.6 EUR
ICNA0215831701 ICNA0215831705
Zincon sodium salt, braunes Pulver
Zincon sodium salt

Zincon is a colorimetric indicator for Zn, Cu and Hg. Reagent for the spectrophotometric determination of zinc. Simultaneous determination of zinc and copper. Used in automated flow injection analysis of zinc. As a metal indicator for EDTA titration of Zn, or for the back titration of Ca in the presence of Mg with glycoletherdiaminetertraacetic acid (GEDTA or EGTA).


Reacts with Zn (pH 8,5 to 9,5) and Cu (pH 5,0 to 9,5) to form blue chelates. Also color reaction occurs with Co, Ni and some other polyvalent cations, Beer's law is observed in the range of 0,1 to 2,4 μg Cu/ml in water at 600 mu.


Slightly soluble in water and alcohol. Insoluble in most organic solvents. Soluble in aqueous alkali resulting in a red solution.

Formel: C₂₀H₁₅N₄NaO₆S
Molecular Weight: 462.412 g/mol
Schmelzpunkt: 250…260 °C (Zers., Lit.)
MDL: MFCD00064385
CAS-Nummer: 62625-22-3

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Storage Store at Room Temperature (15 - 30 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR