cis-4,7,10,13,16,19-Docosahexaensäure ≥98%, farblose Flüssigkeit

Lieferant: MP Biomedicals

Synonyme: DHA, Docosa-4Z,7Z,10Z,13Z,16Z,19Z-Hexaenoic Acid

0215909780
ICNA0215909780EA 197 EUR
ICNA0215909780
cis-4,7,10,13,16,19-Docosahexaensäure ≥98%, farblose Flüssigkeit
cis-4,7,10,13,16,19-Docosahexaenoic acid

Docosahexaenoic acid, DHA, is an omega-3 polyunsaturated fatty acid with 22 carbons and six double bonds, the first double bond occuring at position three from the methyl terminus (22:6; n-3).


Cis-4,7,10,13,16,19-Docosahexaenoic Acid reduces amyloid β production whose accumulation has been found to be associated with the risk of alzheimers maternal docosahexaenoic acid supplementation has been found to decrease lung inflammation in hyperoxia-exposed newborn mice. It also has potent antiinflammatory properties.


DHA is a component of lipid membranes and the myelin sheath. DHA also serves as a precursor for signaling molecules such as prostaglandins and eicosanoids.


Soluble in ethanol, hexane and methylene chloride.

Formel: C₂₂H₃₂O₂
Molecular Weight: 328,50 g/mol
Flammpunkt: 62 °C (143,6 °F) (Lit.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00065722
CAS-Nummer: 6217-54-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR