3-Methylcyclopentadecanon ≥95%, Flüssigkeit

Lieferant: MP Biomedicals

0215918690 0215918680 0215918683
ICNA0215918690EA 483 EUR
ICNA0215918690 ICNA0215918680 ICNA0215918683
3-Methylcyclopentadecanon ≥95%, Flüssigkeit
3-Methylcyclopentadecanon

Insoluble in water. Soluble in ethanol (25 mg/ml) and acetone.

Formel: C₁₆H₃₀O
MDL: MFCD00211114
CAS-Nummer: 541-91-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR