Diphenylcyclopropenon ≥98%, bräunliches Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
1,2-Diphenylcyclopropen-3-on, 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-on
0215952301
ICNA0215952301EA
60.9
EUR
ICNA0215952301
Diphenylcyclopropenon ≥98%, bräunliches Pulver
Diphenylcyclopropenone
Diphenylcyclopropenone is a potent hapten. Its vibration frequency has been evaluated.
Diphenylcyclopropenone was used in topical immunotherapy.
Soluble in acetone; Insoluble in water.
Formel:
C₁₅H₁₀O Molecular Weight: 206,24 g/mol Siedepunkt: 407 °C (1013 hPa) Schmelzpunkt: 113…123 °C Dichte: 1,1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001311 CAS-Nummer: 886-38-4 EINECS: 212-948-4 Merck Index: 12,03366 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...