Micrococcus lysodeikticus, MP Biomedicals
Lieferant: MP Biomedicals
0215997201
ICNA0215997201EA
40.2
EUR
ICNA0215997201
Micrococcus lysodeikticus, MP Biomedicals
Chromatographie-Sorbentien
HPLC-Sorbentien
Lysozyme lysates harvested from cultures of Micrococcus lysodeikticus were attached to sepharose and used for affinity chromatography to isolate various bacteriolytic enzymes and it has also been used in a study to investigate the encapsulation of protein drugs in microparticles.