Ficoll®, MP Biomedicals
Lieferant: MP Biomedicals
0216000380
ICNA0216000380EA 659 EUR
ICNA0216000380
Ficoll®, MP Biomedicals
Zelltrennmedien
A copolymer of sucrose and epichlorohydrin. Component used to make density gradients for lymphocyte separation.