β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂) ≥98%

Lieferant: MP Biomedicals

0216005090 0216005094 0216005050 0216005080 0216005083 0216005091
ICNA0216005090EA 142 EUR
ICNA0216005090 ICNA0216005094 ICNA0216005050 ICNA0216005080 ICNA0216005083 ICNA0216005091
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂) ≥98%
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na2)

White Powder, soluble in water or aqueous buffers (50 mg/ml); soluble in 0,01 N Sodium hydroxide (100 mg/ml - clear to slightly hazy, yellowish solution).


A high purity preparation of NADH suitable for most applications.


Meets or exceeds NRC specifications


β-NADH a pyridine nucleotide and biologically active form of nicotinic acid, is a coenzyme necessary for the catalytic reaction of certain enzymes.


β-Nicotinamide adenine dinucleotide and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalysed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases). As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.


NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, beta-oxidation and the citric acid cycle (Krebs cycle, TCA cycle). It participates in cell signaling events as well, for example as a substrate for the poly (ADP-ribose) polymerases (PARPs) during the DNA damage response. The NAD+/NADH dependent sirtuins play key roles in stress responses during events involving energy metabolism, with implications in cancer biology, diabetes and neurodegenerative disease.


Bitte beachten pH 7,5 - 9,5 (10% aq soln)
Store at −0 °C. Store desiccated. Protect from light. Store under nitrogen.

Formel: C₂₁H₂₇N₇Na₂O₁₄P₂
MDL: MFCD00036200
CAS-Nummer: 606-68-8

Bestellen

Spezifikation Testergebnisse

Synonyms β-NADH, 2Na; Reduced DPN; Cozymase, reduced form; Dihydrophosphopyridine nucleotide; Dihydronicotinamide adenine dinucleotide; Diphosphopyridine nucleotide, reduced form; β-DPNH; DPNH; NADH
Form White Powder
Storage Store at −0 °C. Store desiccated. Protect from light. Store under nitrogen.
pH 7,5 - 9,5 (10% aq soln)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR