N-Lauroylsarkosin ~95%, weißer wachsartiger Feststoff
Lieferant: MP Biomedicals
Synonyme:
N-Dodecanoyl-N-methylglycin
N-Lauroylsarcosine is an anionic detergent.
N-Lauroylsarcosine aids in the solubilization and separation of membrane proteins and glycoproteins. It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. It inhibits hexokinase. It Inhibits bacterial flora of human saliva/gut at 0.25% as well as act as fungistatic agent in aqueous dispersion (1%). It is Useful in concentrated salt solutions, used in the cell lysis step during RNA purification (helps avoid excessive foaming).
Typical Working Concentration: Inhibition of bacterial flora of human saliva/gut=0.25%; fungistatic agent in aqueous dispersion=1%
Soluble in methanol (50 mg/ml - clear, colorless solution).
Formel:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COOH Molecular Weight: 271,4 g/mol Schmelzpunkt: 45…46 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021749 CAS-Nummer: 97-78-9 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ~95% |
FTIR | Conforms to Standard |
Melting Point | Report Result |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...