Amoxicillin
Lieferant: MP Biomedicals
Amoxycillin is an antibacterial agent against gram-positive and gram-negative bacteria.
Amoxicillin is a moderate-spectrum, β-lactam antibiotic. Amoxicillin is one of the most common antibiotics prescribed for children. It is used to study the oxygen dependent antimicrobial systems of polymorphonuclear leukocytes (PMNLs), the risk of resistance development in Helicobacter pylori, and various dosing strategies against Streptococcus pneumoniae and Pneumococcal pneumonia. It is also used to study the synthesis of bacterial cell walls at the level of peptidoglycan polymer chain cross-linking involving bacterial transpeptidase. Amoxicillin is used to study the synthesis of bacterial cell walls at the level of peptidoglycan polymer chain cross-linking involving bacterial transpeptidase.
Amoxicillin inhibits the synthesis of bacterial cell walls. It inhibits cross-linkage between the linear peptidoglycan polymer chains that make up a major component of the cell walls of both gram-positive and gram-negative bacteria.
Soluble in water (Lit.). Typically used at a concentration of 100 ug/ml and is stable in media at 37 °C for approximately 3 days.
Formel:
C₁₆H₁₉N₃O₅S |
MDL:
MFCD00056860 CAS-Nummer: 26787-78-0 |
Spezifikation Testergebnisse
Form | White Crystalline Powder |
Storage | Store at +4 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...