Erythrosin B Dinatriumsalz, rotes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: C.I. 45430, Erythrosin 239

0219044905 0219044910 0219044925
ICNA0219044905EA 18.2 EUR
ICNA0219044905 ICNA0219044910 ICNA0219044925
Erythrosin B Dinatriumsalz, rotes Pulver
Erythrosin B Dinatriumsalz

A red, anionic xanthene dye, has been used as a contrast or counter stain to hematoxylin, crystal violet and other nuclear stains, in both plant and animal tissue.
Erythrosin B Sodium Salt is used as a counter stain to hematoxylin, crystal violet and other nuclear stains, in both plant and animal tissue. Erythosin B dye solution in 0.9% saline is made for intravenous injection to induce occlusion.


  • Concentration: 0.91
  • Soluble in water (20 mg/mL) or ethanol (20 mg/mL)
  • Room Temperature, desiccate

Formel: C₂₀H₆I₄Na₂O₅
Molecular Weight: 879.86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00144257
CAS-Nummer: 16423-68-0

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
UV/VIS: l max (water) 524-528nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR