Natriumdodecylsulfat (SDS) ≥95%

Lieferant: MP Biomedicals
Gefahr

0219052280 0219052290 0219052291
ICNA0219052280EA 25 EUR
ICNA0219052280 ICNA0219052290 ICNA0219052291
Natriumdodecylsulfat (SDS) ≥95%
Natriumdodecylsulfat (SDS)

Soluble in water (200 mg/ml - clear, faint yellow solution), and ethanol (0,1g/10 ml)


SDS is an detergent typically used to solubilise and denature proteins for electrophoresis. It has also been used in large-scale phenol extraction of RNA to promote the dissociation of protein from nucleic acids when extracting from biological material. It is used in Protein Electrophoresis & Blotting.

Formel: CH₃(CH₂)₁₁OSO₃Na
Schmelzpunkt: 204…207 °C
Dichte: 1,1 g/cm³ (20 °C)
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036175
CAS-Nummer: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Synonyms Sodium Lauryl sulphate; sulphuric Acid, Monododecyl Ester, Sodium Salt; Dodecyl sulphate, sodium salt; SLS; SDS; Neutrazyme; Lauryl sulphate Sodium Salt; SDS Running Buffer
Form White Powder
Storage Store at room temperature (15 to 30 °C)
Aggregation Number 62
CMC 8,27 mM
pH 6 - 7 (1% aq soln)
Refractive Index 1,461 (Lit.)
Typical Working Concentration 10 mg SDS/mg protein


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR