Clindamycin hydrochloride

Lieferant: MP Biomedicals

0219116850 0219116810 0219116880
ICNA0219116850EA 394 EUR
ICNA0219116850 ICNA0219116810 ICNA0219116880
Clindamycin hydrochloride
Clindamycin hydrochloride

Synonyms: 7[S]-Chloro-7-deoxylincomycin; Cleocin Hydrochloride


Storage temperature.: +4 °C

Formel: C₁₈H₃₃ClN₂O₅S·HCl
MDL: MFCD07793327
CAS-Nummer: 21462-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR