Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) ≥90%, weißes Pulver

Lieferant: MP Biomedicals

0219123090 0219123091 0219123080 0219123083
ICNA0219123090EA 142 EUR
ICNA0219123090 ICNA0219123091 ICNA0219123080 ICNA0219123083
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) ≥90%, weißes Pulver
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)

Energy-rich precursor in the enzymatic biosynthesis of RNA. A potent vasodilator. Induces contractile responses in some tissues.


Soluble in water (50 mg/ml - clear colourless solution).

Formel: C₉H₁₂N₂Na₃O₁₅P₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00044310
CAS-Nummer: 19817-92-6
EINECS: 243-347-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥90%
OD250/OD260 0,75 ± 0,05
OD280/OD260 0,40 ± 0,05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR